Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Benzimidazolyl)acetonitrile 98%

Supplier: AMBEED, INC
Warning

A309820-25G A309820-100G A309820-10G A309820-500G A309820-5G
76838-558EA 21 USD InStock
76838-558 77263-178 77263-180 77263-182 77263-184
(2-Benzimidazolyl)acetonitrile 98%
(2-Benzimidazolyl)acetonitrile
Precaución: for Research Use Only


Formula: C₉H₇N₃
MDL Number: MFCD00005601
CAS Number: 4414-88-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty