Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromophenyl phenyl ether (BDE No. 3) 98%

Supplier: AMBEED, INC
Danger

A322523-25G A322523-10G A322523-5G A322523-100G
76839-160EA 17 USD InStock
76839-160 77290-306 77290-308 77748-633
4-Bromophenyl phenyl ether (BDE No. 3) 98%
4-Bromophenyl phenyl ether (BDE No. 3)
Precaución: for Research Use Only


Formula: C₁₂H₉BrO
MDL Number: MFCD00000094
CAS Number: 101-55-3
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty