Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sulfabenzamide 98%

Supplier: AMBEED, INC
Warning

A380447-25G A380447-100G A380447-5G
76841-026EA 38 USD InStock
76841-026 77240-382 77240-384 77240-386 77240-388
Sulfabenzamide 98%
Sulfabenzamide
Precaución: for Research Use Only


Formula: C₁₃H₁₂N₂O₃S
MDL Number: MFCD00044890
CAS Number: 127-71-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty