Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-1-pentanol 95%

Supplier: AMBEED, INC
Warning

A422894-25G A422894-100G A422894-10G A422894-5G A422894-500G
76842-110EA 26 USD InStock
76842-110 76979-256 76979-258 76979-260 77673-034
5-Chloro-1-pentanol 95%
5-Chloro-1-pentanol
Precaución: For research use only


Formula: C₅H₁₁ClO
MW: 122.59 g/mol
MDL Number: MFCD00039556
CAS Number: 5259-98-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty