Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methylanthranil acid 98%

Supplier: AMBEED, INC
Warning

A678164-25G A678164-1000G A678164-100G A678164-10G A678164-500G
76844-728EA 6 USD InStock
76844-728 77254-962 77254-964 77254-966 77254-968
5-Methylanthranil acid 98%
5-Methylanthranil acid
Precaución: for Research Use Only


Formula: C₈H₉NO₂
MDL Number: MFCD00007909
CAS Number: 2941-78-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty