Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-6-methylpyridin-3-ol 95%

Supplier: AMBEED, INC
Warning

A762082-1G A762082-100MG A762082-250MG
76845-120EA 256 USD InStock
76845-120 77118-226 77118-228
4-Bromo-6-methylpyridin-3-ol 95%
4-Bromo-6-methylpyridin-3-ol
Precaución: for Research Use Only


Formula: C₆H₆BrNO
MDL Number: MFCD18257655
CAS Number: 1256811-26-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty