Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Aminomethyl)-3,5-difluoropyridine dihydrochloride 98%

Supplier: AMBEED, INC
Warning

A783222-1G A783222-100MG A783222-250MG
76845-246EA 175 USD InStock
76845-246 77159-084 77159-086
2-(Aminomethyl)-3,5-difluoropyridine dihydrochloride 98%
2-(Aminomethyl)-3,5-difluoropyridine dihydrochloride
Precaución: for Research Use Only


Formula: C₆H₆F₂N₂·2ClH
MDL Number: MFCD12827699
CAS Number: 1204298-48-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty