Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(6-Chloropyridin-3-yl)cyclopropanecarboxamide 95%

Supplier: AMBEED, INC
Warning

A818204-1G A818204-100MG A818204-250MG
76845-356EA 186 USD InStock
76845-356 77105-702 77105-704
1-(6-Chloropyridin-3-yl)cyclopropanecarboxamide 95%
1-(6-Chloropyridin-3-yl)cyclopropanecarboxamide
Precaución: for Research Use Only


Formula: C₉H₉ClN₂O
MDL Number: MFCD20702009
CAS Number: 854267-91-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty