Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,6,7,8-Tetrahydronaphthalene-1-carbaldehyde 95%

Supplier: AMBEED, INC
Warning

A104746-100MG A104746-1G A104746-250MG A104746-5G
76869-340EA 44 USD InStock
76869-340 76869-342 76869-344 76869-346
5,6,7,8-Tetrahydronaphthalene-1-carbaldehyde 95%
5,6,7,8-Tetrahydronaphthalene-1-carbaldehyde
Precaución: for Research Use Only


Formula: C₁₁H₁₂O
MDL Number: MFCD15474839
CAS Number: 41828-13-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty