Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4-Chlorophenyl)propiolic acid 98%

Supplier: AMBEED, INC
Warning

A105147-100MG A105147-1G A105147-250MG A105147-5G
76869-384EA 11 USD InStock
76869-384 76869-386 76869-388 76869-390
3-(4-Chlorophenyl)propiolic acid 98%
3-(4-Chlorophenyl)propiolic acid
Precaución: for Research Use Only


Formula: C₉H₅ClO₂
MW: 180.5878 g/mol
MDL Number: MFCD00046550
CAS Number: 3240-10-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty