Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5'-Bromo-2'-chloroacetophenone 98%

Supplier: AMBEED, INC
Warning

A106021-1G A106021-250MG A106021-25G A106021-5G
76869-424EA 8 USD InStock
76869-424 76869-426 76869-428 76869-430
5'-Bromo-2'-chloroacetophenone 98%
5'-Bromo-2'-chloroacetophenone
Precaución: for Research Use Only


Formula: C₈H₆BrClO
MW: 233.4900 g/mol
MDL Number: MFCD02683851
CAS Number: 105884-19-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty