Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fluvastatin sodium salt 98%

Supplier: AMBEED, INC
Warning

A109438-100MG A109438-1MG A109438-250MG A109438-50MG
76869-636EA 20 USD InStock
76869-636 76869-638 76869-640 76869-642
Fluvastatin sodium salt 98%
Fluvastatin sodium salt
Precaución: for Research Use Only


Formula: C₂₄H₂₅FNNaO₄
MW: 433.46 g/mol
MDL Number: MFCD00929076
CAS Number: 93957-55-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty