Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

H-D-Asp(OtBu)-OtBu·HCl 95%

Supplier: AMBEED, INC
Warning

A110197-100G A110197-1G A110197-250MG A110197-25G A110197-5G
76869-672EA 258 USD InStock
76869-672 76869-674 76869-676 76869-678 76869-680
H-D-Asp(OtBu)-OtBu·HCl 95%
H-D-Asp(OtBu)-OtBu·HCl
Precaución: for Research Use Only


Formula: C₁₂H₂₃NO₄·HCl
MDL Number: MFCD09263345
CAS Number: 135904-71-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty