Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-Propylene carbonate 98%

Supplier: AMBEED, INC
Warning

A114110-100G A114110-1G A114110-25G A114110-5G
76869-902EA 145 USD InStock
76869-902 76869-904 76869-906 76869-908
(S)-Propylene carbonate 98%
(S)-Propylene carbonate
Precaución: for Research Use Only


Formula: C₄H₆O₃
MDL Number: MFCD00798264
CAS Number: 51260-39-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty