Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-1-methyl-1H-pyrazole dihydrochloride 97%

Supplier: AMBEED, INC
Warning

A114149-100G A114149-10G A114149-1G A114149-25G A114149-5G
76869-910EA 184 USD InStock
76869-910 76869-912 76869-914 76869-916 76869-918
4-Amino-1-methyl-1H-pyrazole dihydrochloride 97%
4-Amino-1-methyl-1H-pyrazole dihydrochloride
Precaución: for Research Use Only


Formula: C₄H₉Cl₂N₃
MDL Number: MFCD09025437
CAS Number: 1063734-49-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty