Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Chloromethyl)pyrazine hydrochloride 97%

Supplier: AMBEED, INC
Danger

A114708-100MG A114708-1G A114708-5G
76869-944EA 12 USD InStock
76869-944 76869-946 76869-948
2-(Chloromethyl)pyrazine hydrochloride 97%
2-(Chloromethyl)pyrazine hydrochloride


Formula: C₅H₆Cl₂N₂
MDL Number: MFCD16988430
CAS Number: 210037-98-2
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty