Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-4-chloro-3-formylpyridin-2-ylcarbamate 97%

Supplier: AMBEED, INC
Warning

A114954-100MG A114954-250MG
76869-956EA 8 USD InStock
76869-956 76869-958
tert-Butyl-4-chloro-3-formylpyridin-2-ylcarbamate 97%
tert-Butyl-4-chloro-3-formylpyridin-2-ylcarbamate
Precaución: For research use only.


Formula: C₁₁H₁₃ClN₂O₃
MDL Number: MFCD15530295
CAS Number: 868736-42-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty