Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Demeclocycline hydrochloride 90%+ (> 900ug/mg)

Supplier: AMBEED, INC
Warning

A115695-1MG A115695-25MG
76869-986EA 5 USD InStock
76869-986 76869-988
Demeclocycline hydrochloride 90%+ (> 900ug/mg)
Demeclocycline hydrochloride
Precaución: for Research Use Only


Formula: C₂₁H₂₂Cl₂N₂O₈
MW: 501.313 g/mol
MDL Number: MFCD00082371
CAS Number: 64-73-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty