Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Amino-2-fluorophenyl)-4-methylpiperazine 98%

Supplier: AMBEED, INC
Warning

A116215-100MG A116215-1G A116215-250MG A116215-25G A116215-5G
76870-002EA 5 USD InStock
76870-002 76870-004 76870-006 76870-008 76870-010
1-(4-Amino-2-fluorophenyl)-4-methylpiperazine 98%
1-(4-Amino-2-fluorophenyl)-4-methylpiperazine
Precaución: for Research Use Only


Formula: C₁₁H₁₆FN₃
MDL Number: MFCD05663793
CAS Number: 221198-99-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty