Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-5-fluoro-2-nitroaniline 95%

Supplier: AMBEED, INC
Warning

A116462-10G A116462-1G A116462-25G A116462-5G
76870-020EA 137 USD InStock
76870-020 76870-022 76870-024 76870-026
4-Bromo-5-fluoro-2-nitroaniline 95%
4-Bromo-5-fluoro-2-nitroaniline
Precaución: for Research Use Only


Formula: C₆H₄BrFN₂O₂
MDL Number: MFCD11101433
CAS Number: 153505-36-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty