Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Furoic acid 95%

Supplier: AMBEED, INC
Warning

A117549-100G A117549-25G A117549-500G
76870-070EA 6 USD InStock
76870-070 76870-072 76870-074
2-Furoic acid 95%
2-Furoic acid
Precaución: For research use only.


Formula: C₅H₄O₃
MW: 112.08 g/mol
MDL Number: MFCD00003238
CAS Number: 88-14-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty