Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Chloro-4-(piperazin-1-yl)quinoline 95%

Supplier: AMBEED, INC
Warning

A120163-100MG A120163-10G A120163-250MG A120163-5G A120163-1G
76870-176EA 5 USD InStock
76870-176 76870-178 76870-180 76870-182 77670-704
7-Chloro-4-(piperazin-1-yl)quinoline 95%
7-Chloro-4-(piperazin-1-yl)quinoline
Precaución: for Research Use Only


Formula: C₁₃H₁₄ClN₃
MDL Number: MFCD01476123
CAS Number: 837-52-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty