Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-5,6,7,8-tetrahydroquinoline 95%

Supplier: AMBEED, INC
Warning

A120518-100MG A120518-1G A120518-250MG A120518-25MG
76870-184EA 121 USD InStock
76870-184 76870-186 76870-188 76870-190
2,4-Dichloro-5,6,7,8-tetrahydroquinoline 95%
2,4-Dichloro-5,6,7,8-tetrahydroquinoline
Precaución: for Research Use Only


Formula: C₉H₉Cl₂N
MDL Number: MFCD23135262
CAS Number: 858279-01-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty