Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Boc-2,5-diazabicyclo(2.2.2)octane 98%

Supplier: AMBEED, INC
Warning

A120867-100MG A120867-1G A120867-250MG
76870-214EA 78 USD InStock
76870-214 76870-216 76870-218
2-Boc-2,5-diazabicyclo(2.2.2)octane 98%
2-Boc-2,5-diazabicyclo(2.2.2)octane
Precaución: for Research Use Only


Formula: C₁₁H₂₀N₂O₂
MDL Number: MFCD18249863
CAS Number: 858671-91-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty