Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3-(butylcarbamoyl)phenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A123677-1G A123677-250MG A123677-5G
76870-380EA 162 USD InStock
76870-380 76870-382 76870-384
4-Chloro-3-(butylcarbamoyl)phenylboronic acid 98%
4-Chloro-3-(butylcarbamoyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₅BClNO₃
MW: 255.5100 g/mol
MDL Number: MFCD07783886
CAS Number: 871332-94-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty