Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-nitrophenol 95%

Supplier: AMBEED, INC
Danger

A124159-10G A124159-1G A124159-250MG A124159-25G A124159-5G
76870-408EA 177 USD InStock
76870-408 76870-410 76870-412 76870-414 76870-416
3-Chloro-2-nitrophenol 95%
3-Chloro-2-nitrophenol
Precaución: for Research Use Only


Formula: C₆H₄ClNO₃
MDL Number: MFCD08703192
CAS Number: 17802-02-7
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty