Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-2,5-dichloropyridine 98%

Supplier: AMBEED, INC
Warning

A124816-100MG A124816-1G A124816-5G
76870-432EA 7 USD InStock
76870-432 76870-434 76870-436
4-Amino-2,5-dichloropyridine 98%
4-Amino-2,5-dichloropyridine
Precaución: for Research Use Only


Formula: C₅H₄Cl₂N₂
MDL Number: MFCD11044256
CAS Number: 405230-82-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty