Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Methylsulfonyl)propyl methanesulfonate 95%

Supplier: AMBEED, INC
Warning

A126721-100MG A126721-250MG
76870-544EA 14 USD InStock
76870-544 76870-546
3-(Methylsulfonyl)propyl methanesulfonate 95%
3-(Methylsulfonyl)propyl methanesulfonate
Precaución: for Research Use Only


Formula: C₅H₁₂O₅S₂
MDL Number: MFCD18642728
CAS Number: 357913-53-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty