Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-[(1S)-1-aminoethyl]benzoate 98%

Supplier: AMBEED, INC
Warning

A127228-10G A127228-1G A127228-250MG A127228-25G A127228-5G
76870-570EA 97 USD InStock
76870-570 76870-572 76870-574 76870-576 76870-578
Methyl 4-[(1S)-1-aminoethyl]benzoate 98%
Methyl 4-[(1S)-1-aminoethyl]benzoate
Precaución: for Research Use Only


Formula: C₁₀H₁₃NO₂
MW: 179.216 g/mol
MDL Number: MFCD06762070
CAS Number: 222714-37-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty