Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Enrofloxacin 98+%

Supplier: AMBEED, INC
Warning

A127543-10G A127543-1G A127543-1MG A127543-5G A127543-100G
76870-590EA 22 USD InStock
76870-590 76870-592 76870-594 76870-596 77611-100
Enrofloxacin 98+%
Enrofloxacin
Precaución: for Research Use Only


Formula: C₁₉H₂₂FN₃O₃
MDL Number: MFCD00792463
CAS Number: 93106-60-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty