Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-8-methylquinoline 98%

Supplier: AMBEED, INC
Warning

A130764-1G A130764-250MG A130764-5G
76870-708EA 331.01 USD InStock
76870-708 76870-710 76870-712 76870-714
2-Chloro-8-methylquinoline 98%
2-Chloro-8-methylquinoline
Precaución: for Research Use Only


Formula: C₁₀H₈ClN
MDL Number: MFCD11108682
CAS Number: 4225-85-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty