Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-(-)-α-Phenylglycine 98%

Supplier: AMBEED, INC
Warning

A132525-1000G A132525-100G A132525-25G A132525-500G
76870-806EA 133 USD InStock
76870-806 76870-808 76870-810 76870-812
D-(-)-α-Phenylglycine 98%
D-(-)-α-Phenylglycine
Precaución: for Research Use Only


Formula: C₈H₉NO₂
MW: 151.16 g/mol
MDL Number: MFCD00008061
CAS Number: 875-74-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty