Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-N-(5-chloropyridin-2-yl)-5-methoxybenzamide 98%

Supplier: AMBEED, INC
Warning

A134499-10G A134499-1G A134499-5G
76870-898EA 62 USD InStock
76870-898 76870-900 76870-902 76870-904
2-Amino-N-(5-chloropyridin-2-yl)-5-methoxybenzamide 98%
2-Amino-N-(5-chloropyridin-2-yl)-5-methoxybenzamide
Precaución: for Research Use Only


Formula: C₁₃H₁₂ClN₃O₂
MW: 277.71 g/mol
MDL Number: MFCD12841614
CAS Number: 280773-17-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty