Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-Oxo-4,7-dihydro-1H-pyrrolo[2,3-d]pyrimidin-2-yl)pivalamide 97%

Supplier: AMBEED, INC
Warning

A134558-1G A134558-250MG A134558-5G
76870-906EA 562.28 USD InStock
76870-906 76870-908 76870-910 76870-912
N-(4-Oxo-4,7-dihydro-1H-pyrrolo[2,3-d]pyrimidin-2-yl)pivalamide 97%
N-(4-Oxo-4,7-dihydro-1H-pyrrolo[2,3-d]pyrimidin-2-yl)pivalamide
Precaución: for Research Use Only


Formula: C₁₁H₁₄N₄O₂
MDL Number: MFCD11109810
CAS Number: 137281-08-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty