Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dibromo-4-chloropyridine 95%

Supplier: AMBEED, INC
Warning

A134786-10G A134786-1G A134786-250MG A134786-5G
76870-930EA 237 USD InStock
76870-930 76870-932 76870-934 76870-936
2,6-Dibromo-4-chloropyridine 95%
2,6-Dibromo-4-chloropyridine
Precaución: for Research Use Only


Formula: C₅H₂Br₂ClN
MDL Number: MFCD13189630
CAS Number: 1196156-59-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty