Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Chloromethyl)thiazole hydrochloride 97%

Supplier: AMBEED, INC
Danger

A137297-1G A137297-250MG A137297-100MG
76871-010EA 73 USD InStock
76871-010 76871-012 77670-968
5-(Chloromethyl)thiazole hydrochloride 97%
5-(Chloromethyl)thiazole hydrochloride
Precaución: for Research Use Only


Formula: C₄H₅Cl₂NS
MDL Number: MFCD22380404
CAS Number: 131052-44-3
UN: 1759
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty