Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Fluorophenyl)cyclohexanecarboxylic acid 95%

Supplier: AMBEED, INC
Warning

A138702-1G A138702-250MG
76871-086EA 55 USD InStock
76871-086 76871-088
1-(2-Fluorophenyl)cyclohexanecarboxylic acid 95%
1-(2-Fluorophenyl)cyclohexanecarboxylic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₅FO₂
MDL Number: MFCD00800623
CAS Number: 106795-66-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty