Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Benzyloxyindole-3-carbaldehyde 97%

Supplier: AMBEED, INC
Warning

A138813-10G A138813-1G A138813-250MG A138813-5G
76871-094EA 97 USD InStock
76871-094 76871-096 76871-098 76871-100
6-Benzyloxyindole-3-carbaldehyde 97%
6-Benzyloxyindole-3-carbaldehyde
Precaución: for Research Use Only


Formula: C₁₆H₁₃NO₂
MDL Number: MFCD00056931
CAS Number: 92855-64-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty