Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-5-chloroindole-3-boronic acid pinacol ester 95%

Supplier: AMBEED, INC
Warning

A139055-100MG A139055-1G A139055-250MG A139055-5G
76871-114EA 30 USD InStock
76871-114 76871-116 76871-118 76871-120
1-Boc-5-chloroindole-3-boronic acid pinacol ester 95%
1-Boc-5-chloroindole-3-boronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₉H₂₅BClNO₄
MDL Number: MFCD15143612
CAS Number: 1218790-30-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty