Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3,5-xylenol 98%

Supplier: AMBEED, INC
Warning

A139376-1000G A139376-100G A139376-25G A139376-500G
76871-130EA 55 USD InStock
76871-130 76871-132 76871-134 76871-136 76871-138
4-Chloro-3,5-xylenol 98%
4-Chloro-3,5-xylenol
Precaución: for Research Use Only


Formula: C₈H₉ClO
MDL Number: MFCD00002324
CAS Number: 88-04-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty