Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Benzyloxy-6-azaindole 95%

Supplier: AMBEED, INC
Warning

A143244-100MG A143244-1G A143244-250MG
76871-304EA 11 USD InStock
76871-304 76871-306 76871-308
5-Benzyloxy-6-azaindole 95%
5-Benzyloxy-6-azaindole
Precaución: for Research Use Only


Formula: C₁₄H₁₂N₂O
MW: 224.258 g/mol
MDL Number: MFCD09038914
CAS Number: 17288-54-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty