Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-4-(isobutylsulfonyl)benzene 97%

Supplier: AMBEED, INC
Warning

A144307-1G A144307-250MG A144307-5G
76871-368EA 147 USD InStock
76871-368 76871-370 76871-372
1-Bromo-4-(isobutylsulfonyl)benzene 97%
1-Bromo-4-(isobutylsulfonyl)benzene
Precaución: for Research Use Only


Formula: C₁₀H₁₃BrO₂S
MDL Number: MFCD20441890
CAS Number: 856060-51-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty