Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-1-(Pyrrolidin-3-yl)piperidine dihydrochloride 95%

Supplier: AMBEED, INC
Warning

A145781-100MG A145781-1G A145781-250MG
76871-440EA 34 USD InStock
76871-440 76871-442 76871-444
(S)-1-(Pyrrolidin-3-yl)piperidine dihydrochloride 95%
(S)-1-(Pyrrolidin-3-yl)piperidine dihydrochloride
Precaución: for Research Use Only


Formula: C₉H₂₀Cl₂N₂
MDL Number: MFCD11849811
CAS Number: 917505-12-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty