Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-(tert-butyl)pyridine 97%

Supplier: AMBEED, INC
Warning

A148172-100MG A148172-25G A148172-5G
76871-508EA 13 USD InStock
76871-508 76871-510 76871-512
2-Bromo-4-(tert-butyl)pyridine 97%
2-Bromo-4-(tert-butyl)pyridine
Precaución: for Research Use Only


Formula: C₉H₁₂BrN
MDL Number: MFCD11223221
CAS Number: 50488-34-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty