Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-2-methyl-D-phenylalanine 97%

Supplier: AMBEED, INC
Warning

A149842-1G A149842-250MG A149842-25G A149842-5G
76871-566EA 19 USD InStock
76871-566 76871-568 76871-570 76871-572
Boc-2-methyl-D-phenylalanine 97%
Boc-2-methyl-D-phenylalanine
Precaución: for Research Use Only


Formula: C₁₅H₂₁NO₄
MW: 279.34 g/mol
MDL Number: MFCD00671403
CAS Number: 80102-29-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty