Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-amino-2-sulfamoylbenzoate 98%

Supplier: AMBEED, INC
Warning

A151739-1G A151739-250MG A151739-5G
76871-620EA 52 USD InStock
76871-620 76871-622 76871-624
Methyl 4-amino-2-sulfamoylbenzoate 98%
Methyl 4-amino-2-sulfamoylbenzoate
Precaución: for Research Use Only


Formula: C₈H₁₀N₂O₄S
MDL Number: MFCD22421606
CAS Number: 2297-06-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty