Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

OSU-03012 98+%

Supplier: AMBEED, INC
Warning

A152748-1MG A152748-5MG
76871-672EA 9 USD InStock
76871-672 76871-674
OSU-03012 98+%
OSU-03012
Precaución: for Research Use Only


Formula: C₂₆H₁₉F₃N₄O
MW: 460.45 g/mol
MDL Number: MFCD12912272
CAS Number: 742112-33-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty