Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-m-toluic acid 98%

Supplier: AMBEED, INC
Warning

A153003-100G A153003-500G A153003-100MG
76871-702EA 28 USD InStock
76871-702 76871-704 77318-266 77671-142
2-Hydroxy-m-toluic acid 98%
2-Hydroxy-m-toluic acid
Precaución: for Research Use Only


Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 293
MDL Number: MFCD00002448
CAS Number: 83-40-9
Merck Index: 13,02609
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty