Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(6-Bromopyridin-3-yl)acrylic acid 98%

Supplier: AMBEED, INC
Warning

A154199-100MG A154199-1G A154199-250MG
76871-778EA 24 USD InStock
76871-778 76871-780 76871-782
3-(6-Bromopyridin-3-yl)acrylic acid 98%
3-(6-Bromopyridin-3-yl)acrylic acid
Precaución: for Research Use Only


Formula: C₈H₆BrNO₂
MW: 228.04 g/mol
MDL Number: MFCD11504804
CAS Number: 1035123-89-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty