Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Naphthoic acid 98%

Supplier: AMBEED, INC
Warning

A155554-100G A155554-10G A155554-25G A155554-500G
76871-832EA 25 USD InStock
76871-832 76871-834 76871-836 76871-838
2-Naphthoic acid 98%
2-Naphthoic acid
Precaución: for Research Use Only


Formula: C₁₁H₈O₂
MW: 172.18 g/mol
Boiling Pt: 333
MDL Number: MFCD00004101
CAS Number: 93-09-4
Merck Index: 12,06469
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty